Screw Caps, MicroSolv™
PP, Tan Silicone Rubber / White PTFE, bonded, 9-425 mm, Blue, Screw cap
Catalog # 97055-306
Supplier: MicroSolv Technology Corp
Fournisseur #: 9502S-11-MB-K
Jump to:
Product Details & Documents
Spécifications
- Taille du bouchon:9-425 mm
- Cap Type:Screw cap
- Septum:Tan Silicone Rubber / White PTFE, bonded
- Matière:PP
- Cap Color:Blue