Headspace Caps, MicroSolv
Magnetic Screw Cap, 18 mm Precision Threads, Closed, Aluminum, Silicone Butyl Red/PTFE Grey, 55° Shore A 1.6 mm Septa, Silver
Catalog # 97054-922
Fournisseur #: 95020-55S-CD
Jump to:
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
MAGNETIC SCRW TOP HEADSPACE VIAL CAPS Catalog #10827-872
Le produit 10827-872 est similaire à 97054-922 mais le fabricant et les spécifications peuvent varier.
Product Details & Documents
MicroSolv™ headspace caps are available as magnetic and with 4 different septa types to meet needs.
The screw cap eliminates the need for heavy crimping tools and assures that the vials are sealed perfectly every time. The design saves time and money, and ensures a good head space analysis.
Spécifications
- Septum:Silicone Butyl Red/PTFE Grey, 55° Shore A 1.6 mm Septa
- Color:Silver
- Désignation:Magnetic Screw Cap
- Matière:Aluminum
- Taille:18 mm Precision Threads, Closed