PreColumn™ SS Filter for HPLC, MicroSolv | Avantor
PreColumn™ SS Filter for HPLC, MicroSolv
PEEK, 2 µm
Catalog # 97054-864
Fournisseur #:  49650-020
PreColumn™ SS Filter for HPLC, MicroSolv
Catalog # 97054-864
PreColumn™ SS Filter for HPLC, MicroSolv
PreColumn™ SS Filter for HPLC, MicroSolv
PEEK, 2 µm
Price
$417.74
Log in pour la disponibilité
Pack of 10
La quantité minimale de commande est de 1
Product Details & Documents
Protect your HPLC columns from particulates that can ruin them at ¹/₃ the cost of conventional precolumn filters.
When back pressure levels increase to unacceptable levels, changes remove the filter quickly to minimize instrument downtime while increasing column life and dispose of the precolumn filter.
The precolumn body contains a stainless steel filter element and does not introduce any extra column volume so your chromatography is not affected.
With its convenient direct connect design, changeover time is measured in seconds and no tools or wrenches are needed.
Ordering information: Use with ¹/₁₆" 10 - 32 internal fitting ports.
Spécifications
  • Taille de pore:
    2 µm
  • Désignation:
    PEEK
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.