Screw Caps, MicroSolv™
PP, Bonded Silicone/PTFE, pre-slit, 9-425 mm, Blue, Screw cap, knurled
Catalog # 97051-428
Supplier: MicroSolv Technology Corp
Fournisseur #: 9502S-30C-B-M
Jump to:
Product Details & Documents
These blue screw caps are designed for use with 9-425, 12x32 mm vials.
Spécifications
- Taille du bouchon:9-425 mm
- Cap Type:Screw cap, knurled
- Septum:Bonded Silicone/PTFE, pre-slit
- Matière:PP
- Cap Color:Blue