Screw Caps, MicroSolv™
PP, Bonded Silicone/PTFE, low durometer, 9-425 mm, Blue, Screw cap, knurled
97051-424
: 9502S-10C-B-M
These blue screw caps are designed for use with 9-425, 12x32 mm vials.
- Taille du bouchon:9-425 mm
- Cap Type:Screw cap, knurled
- Septum:Bonded Silicone/PTFE, low durometer
- Matière:PP
- Cap Color:Blue