Screw-Thread Caps, 9 mm, MicroLiter, DWK Life Sciences
Yellow Polypropylene, FEP/Butyl, 9 mm
  97050-361
Screw-Thread Caps, 9 mm, MicroLiter, DWK Life Sciences
  97050-361
 :  
Screw-Thread Caps, 9 mm, MicroLiter, DWK Life Sciences
Screw-Thread Caps, 9 mm, MicroLiter, DWK Life Sciences
Yellow Polypropylene, FEP/Butyl, 9 mm
MicroLiter's 9 mm screw-thread caps are available in multiple colors and septa types.
  • Ø:
    9 mm
  • Septum:
    FEP/Butyl
  • Désignation:
    Yellow Polypropylene
Complementary Products
  •  
    Bottom Style
    Description
    Capacity
    Material
    Cap included
    Ø
    Septum
    Cap Size
  • 97046-218
    Septa
    9 mm
    Tefzel®/burnt red slicone
    Pack of 100 - $36.48
  • 76045-380
    Non-Slit Caps
    9 mm
    Case of 100 - $82.36
  • 97046-232
    Septa
    9 mm
    PTFE/silicone
    Pack of 100 - $46.75
  • 97046-176
    Septa
    9 mm
    PTFE
    Pack of 100 - $13.68
  • 97046-179
    Septa
    9 mm
    FEP/natural rubber
    Pack of 100 - $38.21
  • 97046-264
    Septa
    9 mm
    Precut PTFE/silicone
    Pack of 100 - $89.95
  • 76045-382
    Slit Caps
    9 mm
    Case of 100 - $87.02
  • 97046-204
    Septa
    9 mm
    FEB/butyl
    Pack of 100 - $32.39
  • 97046-284
    Septa
    9 mm
    PTFE/silicone/PTFE
    Pack of 100 - $71.25