Solid Polyethylene Acid Cabinets, Justrite®
Acids underbench cabinet, 2 door, undercounter, 36× 2½ L bottles, Polyethylene
Catalog # 97048-666
Supplier: Justrite
Supplier #: 24010
Jump to:
Product Details & Documents
These solid wall high density polyethylene cabinets have excellent chemical resistance to handle the most corrosive chemicals.
- Sturdy, smooth construction in an attractive design
- Suitable for highly aggressive liquids - no metal parts to corrode
- Three-point door latch on larger units offer secure closure
- Toe kick on undercounter model allows user to stand closer to counter - less leaning, better ergonomics
They are 100% poly welded with no metal parts, they include a external 2" NPT vent/exhaust connections and have padlockable hasps for security. White.
Spécifications
- Capacité:2 door, undercounter, 36× 2½ L bottles
- Désignation:Acids underbench cabinet
- Matière:Polyethylene
- Cat. No.:97048-666