WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences | Avantor
WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences
Vial, 4.00 ml, Amber, Oui, Open top, phenolic, Black, PTFE
Catalog # 97047-677
WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences
Catalog # 97047-677
Supplier:  
WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences
WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences
Vial, 4.00 ml, Amber, Oui, Open top, phenolic, Black, PTFE
Discontinued
Product Details & Documents
These component kits are shipped with a pack of vials, a pack of caps and a pack of septa.
Available in clear and amber, with polypropylene or phenolic closures and a variety of styles of liners and septa.
Spécifications
  • Avec bouchon:
    Oui
  • Cap Type:
    Open top, phenolic
  • Septum:
    PTFE
  • Capacité:
    4.00 ml
  • Color:
    Amber
  • Désignation:
    Vial
  • Cap Color:
    Black