WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences
Vial, 4.00 ml, Amber, Oui, Open top, phenolic, Black, PTFE
Catalog # 97047-677
Jump to:
Discontinued
Product Details & Documents
These component kits are shipped with a pack of vials, a pack of caps and a pack of septa.
Available in clear and amber, with polypropylene or phenolic closures and a variety of styles of liners and septa.
Spécifications
- Avec bouchon:Oui
- Cap Type:Open top, phenolic
- Septum:PTFE
- Capacité:4.00 ml
- Color:Amber
- Désignation:Vial
- Cap Color:Black