WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences
Vial, 4.00 ml, Clear, Oui, Solid top, phenolic, Black, PTFE
Catalog # 97047-642
 :  13-1010KPS
WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences
Catalog # 97047-642
WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences
WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences
Vial, 4.00 ml, Clear, Oui, Solid top, phenolic, Black, PTFE
Price
$139.65
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 100
Product Details & Documents
These component kits are shipped with a pack of vials, a pack of caps and a pack of septa.
Available in clear and amber, with polypropylene or phenolic closures and a variety of styles of liners and septa.
Spécifications
  • Avec bouchon:
    Oui
  • Cap Type:
    Solid top, phenolic
  • Septum:
    PTFE
  • Capacité:
    4.00 ml
  • Color:
    Clear
  • Désignation:
    Vial
  • Cap Color:
    Black