Screw-Thread Caps, 9 mm, MicroLiter, DWK Life Sciences
Blue Polypropylene, Cut PTFE/Silicone, 9 mm
Supplier: DWK Life Sciences (KIMBLE)
Supplier #: 09-0035B-B
Screw-Thread Caps, 9 mm, MicroLiter, DWK Life Sciences
Catalog # 97046-267
Supplier: DWK Life Sciences (KIMBLE)
Jump to:
Product Details & Documents
MicroLiter's 9 mm screw-thread caps are available in multiple colors and septa types.
Specifications
- Ø:9 mm
- Septum:Cut PTFE/Silicone
- Désignation:Blue Polypropylene
Complementary Products
Product Information
- Catalog #Bottom StyleDescriptionCapacityMaterialCap includedØSeptumCap SizeAvailabilityPrice