WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences
Vial with patch, 2.00 ml, Clear, Oui, Solid top, Black, PTFE/silicone
Catalog # 97046-119
WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences
Catalog # 97046-119
Supplier:  
WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences
WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences
Vial with patch, 2.00 ml, Clear, Oui, Solid top, Black, PTFE/silicone
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
VIAL 2ML 8MM WPTFESIL Y PK100 Catalog #66106-242
VIAL KITS W/CAP+SEPTA PK100 Catalog #66008-058
Product Details & Documents
These component kits are shipped with a pack of vials, a pack of caps and a pack of septa.
Available in clear and amber, with polypropylene or phenolic closures and a variety of styles of liners and septa.
Spécifications
  • Avec bouchon:
    Oui
  • Cap Type:
    Solid top
  • Septum:
    PTFE/silicone
  • Capacité:
    2.00 ml
  • Color:
    Clear
  • Désignation:
    Vial with patch
  • Cap Color:
    Black