WHEATON® MicroLiter Sample Vial Component Kits, DWK Life Sciences
Vial with patch, 2.00 ml, Clear, Oui, Solid top, Black, PTFE/silicone
Catalog # 97046-119
Jump to:
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
Product Details & Documents
These component kits are shipped with a pack of vials, a pack of caps and a pack of septa.
Available in clear and amber, with polypropylene or phenolic closures and a variety of styles of liners and septa.
Spécifications
- Avec bouchon:Oui
- Cap Type:Solid top
- Septum:PTFE/silicone
- Capacité:2.00 ml
- Color:Clear
- Désignation:Vial with patch
- Cap Color:Black