Cleaning Solution, Pepsin/Hydrochloric Acid Solution, SI Analytics
Pepsin/Hydrochloric Acid Solution, 250 mL, Glass bottle, DURAN®
Supplier: SI Analytics
Fournisseur #: 285138295
Cleaning Solution, Pepsin/Hydrochloric Acid Solution, SI Analytics
Catalog # 97042-452
Supplier: SI Analytics
Jump to:
Product Details & Documents
Suitable for cleaning combination electrodes and reference electrodes.
Spécifications
- Désignation:Pepsin/Hydrochloric Acid Solution
- Taille:250 mL
- Packaging:Glass bottle, DURAN®