Purifier® Cell Logic Accessories, Labconco®
Product Details & Documents
Purifier Cell Logic Class II Biosafety Cabinets are specially designed with cell researchers in mind, while also providing protection from hazardous particulates such as agents that require Biosafety Level 1,2 or 3 containment.
Scope-Ready and Temp-Zone Packages are available. Many accessories are available for these packages. For example, Aluminum Tube Racks, Blue anodized aluminum blocks that hold tubes upright and promote conduction of heat or cold from the Temp-Zone area to help maintain sample temperature, are available.
The Pure-Vu Seal fits most inverted scopes with a relatively stationary nominal eyepiece height (distance from base of microscope to centerline of eyepiece) of 13". For use with Purifier Cell Logic Biosafety Cabinet with Scope-Ready package. Contact Labconco with questions about additional accessories.
Spécifications
- Désignation:Pure-Vu XL Seal, Scope-Ready Package