Purifier® Cell Logic Accessories, Labconco®
Purifier® Cell Logic Accessories, Labconco®
Pure-Vu Punch Kit, Scope-Ready Package
Catalog # 97041-750
Supplier:   Labconco
Supplier #:  4027801
Purifier® Cell Logic Accessories, Labconco®
Catalog # 97041-750
Supplier:  Labconco
Purifier® Cell Logic Accessories, Labconco®
Purifier® Cell Logic Accessories, Labconco®
Pure-Vu Punch Kit, Scope-Ready Package
Price
$195.00
Log in pour la disponibilité
Each
Minimum order quantity is 1
Product Details & Documents
Purifier Cell Logic Class II Biosafety Cabinets are specially designed with cell researchers in mind, while also providing protection from hazardous particulates such as agents that require Biosafety Level 1,2 or 3 containment.
Scope-Ready and Temp-Zone Packages are available. Many accessories are available for these packages. For example, Aluminum Tube Racks, Blue anodized aluminum blocks that hold tubes upright and promote conduction of heat or cold from the Temp-Zone area to help maintain sample temperature, are available.
The Pure-Vu Seal fits most inverted scopes with a relatively stationary nominal eyepiece height (distance from base of microscope to centerline of eyepiece) of 13". For use with Purifier Cell Logic Biosafety Cabinet with Scope-Ready package. Contact Labconco with questions about additional accessories.
Spécifications
  • Désignation:
    Pure-Vu Punch Kit, Scope-Ready Package