HPLC Columns, YMC-Pack Diol-SEC
Analytique, Diol-SEC, 200 Å, Verre, 5 µm, 8.00 mm, 300 mm, Silica
97030-796
: DL20S05-3008FG
Diol-SEC is a size exclusion chromatography column utilizing a dihydroxypropyl stationary phase bonded to a silica gel base and useful for molecular weight determination of proteins and sugars.
- Silica-based column with high mechanical strength
- Very low nonspecific adsorption of proteins
- More accurate quantitation of monoclonal antibodies with two products
- Provides improvement in resolution and throughput on size exclusion chromatography
Diol-120, 200, and 300 are suitable for separation or molecular weight determination of proteins with molecular weights of 10000 to several hundred thousand. Diol-60 is the most suitable for separation of peptides of oligosaccharides whose molecular weights are 10000 or less.
- Type de colonne:Analytique
- Ø int.:8.00 mm
- Granulométrie:5 µm
- Taille de pore:200 Å
- Longueur:300 mm
- Column Hardware:Verre
- Packing Material:Silica
- Phase:Diol-SEC
- Cat. No.:97030-796
- pH range:5 - 7.5



