HPLC Columns, YMC-Pack Diol-SEC
Colonne de garde, Diol-SEC, 200 Å, Acier inoxydable, 5 µm, 8.00 mm, 30 mm, Silica
Catalog # 97030-790
Supplier: YMC America,Inc
Fournisseur #: DL20S05-0308WTG
Jump to:
Product Details & Documents
Diol-SEC is a size exclusion chromatography column utilizing a dihydroxypropyl stationary phase bonded to a silica gel base and useful for molecular weight determination of proteins and sugars.
- Silica-based column with high mechanical strength
- Very low nonspecific adsorption of proteins
- More accurate quantitation of monoclonal antibodies with two products
- Provides improvement in resolution and throughput on size exclusion chromatography
Diol-120, 200, and 300 are suitable for separation or molecular weight determination of proteins with molecular weights of 10000 to several hundred thousand. Diol-60 is the most suitable for separation of peptides of oligosaccharides whose molecular weights are 10000 or less.
Spécifications
- Type de colonne:Colonne de garde
- Ø int.:8.00 mm
- Granulométrie:5 µm
- Taille de pore:200 Å
- Longueur:30 mm
- Column Hardware:Acier inoxydable
- Packing Material:Silica
- Phase:Diol-SEC
- Cat. No.:97030-790
- pH range:5 - 7.5
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Cartridge Holder, 1.0 – 4.0 mm for 10 mm Length CartridgesCartridge Holder, 1.0 – 4.0 mm for 10 mm Length Cartridges89230-180Log in pour la disponibilitéEach - $664.26



