Accessories for General Purpose Heating Circulators, Grant Instruments
Accessories for General Purpose Heating Circulators, Grant Instruments
Test Tube Rack for 16-19 mm Tubes (36 Places) not for 5 L Tank
Catalog # 97025-608
Supplier:   GRANT USA INC.
Supplier #:  VR-19
Accessories for General Purpose Heating Circulators, Grant Instruments
Catalog # 97025-608
Supplier:  GRANT USA INC.
Accessories for General Purpose Heating Circulators, Grant Instruments
Accessories for General Purpose Heating Circulators, Grant Instruments
Test Tube Rack for 16-19 mm Tubes (36 Places) not for 5 L Tank
Price
$364.98
Log in pour la disponibilité
Each
Product Details & Documents
Spécifications
  • Désignation:
    Test Tube Rack for 16-19 mm Tubes (36 Places) not for 5 L Tank
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Clamp to attach Thermostat to Independent Tank; allows independent use of Optima™ Immersion Thermostat, models T100, TC120, TX150, and TXF200
    Clamp to attach Thermostat to Independent Tank; allows independent use of Optima™ Immersion Thermostat, models T100, TC120, TX150, and TXF200
    89260-854
    Log in pour la disponibilité
    Each - $268.61
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Bath Draining Siphon
    Bath Draining Siphon
    97025-598
    Log in pour la disponibilité
    Each - $120.07
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Spheres, Polypropylene (300)
    Spheres, Polypropylene (300)
    66238-449
    Log in pour la disponibilité
    Pack of 300 - $245.12