HEPA Filter, Labconco® | Avantor
HEPA Filter, Labconco®
Replacement Exhaust HEPA Filter for 5' Purifier Logic
Catalog # 97006-996
Supplier:   Labconco
Fournisseur #:  3838502
HEPA Filter, Labconco®
Catalog # 97006-996
Supplier:  Labconco
HEPA Filter, Labconco®
HEPA Filter, Labconco®
Replacement Exhaust HEPA Filter for 5' Purifier Logic
Price
$2,340.00
Log in pour la disponibilité
Each
Product Details & Documents
Replacement HEPA filter for use with XPert filtered systems and stations, XPert TXE equipment enclosures, FilterMate portable exhausters, Purifier logic biosafety sabinets, Purifier Cell Logic biosafety cabinets and Double HEPA filtration kits.
  • 99.99% efficient on particles 0.3 µm in size
  • Available in 18.0", 24.0", 30.0", and 42.0" widths
Have aluminum frames with either aluminum separators or separatorless pleats.
Ordering information: Note: Units 5' and 6' wide require two filters.
Spécifications
  • Désignation:
    Replacement Exhaust HEPA Filter for 5' Purifier Logic