Temperature Chart Recorders, Dickson®
Temperature Chart Recorders, Dickson®
24-Hour Chart, 7...32 °C
Catalog # 97006-896
Supplier:   Dickson
Supplier #:  C207
Temperature Chart Recorders, Dickson®
Catalog # 97006-896
Supplier:  Dickson
Temperature Chart Recorders, Dickson®
Temperature Chart Recorders, Dickson®
24-Hour Chart, 7...32 °C
Price
$192.29
Log in for availability
Pack of 60
Product Details & Documents
Recorders are ideal for monitoring long-term temperature trends in areas where proper temperatures are critical to product stability.
  • Cord-free, portable, and easy to use
  • Recorders can be placed directly in an environment that needs to be monitored, and mounted on a desk or wall
  • Feature spiral-wound bimetallic strip transducer, hinged door, built-in slot for optional tie-down cable and lock, battery door
  • Durable, corrosion-resistant black ABS and polycarbonate enclosure
  • Recorders come with or without digital LCD, and can be set to 7-day or 24-hour recording times
  • °C/°F switchable
Designed for use in blood banks, laboratories, storage areas, refrigerators, or freezers.
Average response time: 11 min. to move 63% of full scale
Certifications: A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology. CE marked. IP21 rated.
Ordering information: Each recorder comes with a AA alkaline battery and a red pen. Charts must be purchased separately.
Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
Specifications
  • Plage de température:
    7...32 °C
  • Désignation:
    24-Hour Chart
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Red Pens
    Red Pens
    61280-132
    Log in for availability
    Pack of 6 - $244.14
Product Information
  • Catalog #
    Availability
    Price
  • NIST Traceable Calibration 1-Point Service
    NIST Traceable Calibration 1-Point Service
    13450-754
    Log in for availability
    Each - $291.26
Product Information
  • Catalog #
    Availability
    Price
  • NIST & A2LA Traceable Calibration 3-Point Service
    NIST & A2LA Traceable Calibration 3-Point Service
    13450-756
    Log in for availability
    Each - $848.52