EXP® Pre-Column Filters, Optimize Technologies
EXP® Pre-Column Filters, Optimize Technologies
EXP® Pre-column filter cartridge, 0.5 µm, 0.5 µm, 5 replacement cartridges
Catalog # 97004-010
Supplier:   Optimize
Supplier #:  15-04-03093
EXP® Pre-Column Filters, Optimize Technologies
Catalog # 97004-010
Supplier:  Optimize
EXP® Pre-Column Filters, Optimize Technologies
EXP® Pre-Column Filters, Optimize Technologies
EXP® Pre-column filter cartridge, 0.5 µm, 0.5 µm, 5 replacement cartridges
Price
$1,467.03
Log in for availability
Pack of 5
Product Details & Documents
The EXP® Pre-Column Filter is ideal for protecting small-particle UHPLC columns at extreme pressures.
  • Approved for use at 20000+ psi (1400+ bar)
  • Hand-tight filter replacement - no tools required
  • Depth filtration maximizes capacity
  • Uni-directional design
  • Low-volume/low-dispersion cartridges
  • Use with 15-04-03838
Such techniques analyze samples in the most demanding applications, which can decrease the life of these expensive columns. EXP® Pre-Column Filters help extend that life and protect your column investment without sacrificing performance. Free-Turn¨ architecture allows the user to change cartridges by hand without breaking fluid connections on the holder inlet/outlet. EXP® Pre-Column Filter holders are available with Titanium Hybrid ferrules for easy direct connection to any 10-32 port.
Ordering information: Custom volumes/porosities available.
Delivery information: The filter holder comes as a complete package including fittings to provide repeated zero-dead volume column connections.
Specifications
  • Taille de pore:
    0.5 µm
  • Désignation:
    EXP® Pre-column filter cartridge, 0.5 µm
  • Includes:
    5 replacement cartridges