Flexible Vent Assemblies, Scientific Plastics
Double cabinet penetration, Kewaunee* cabinets with Labconco* worsurfaces
Catalog # 97002-256
Supplier:   Scientific Plastics
 :  F160L078F-150
Flexible Vent Assemblies, Scientific Plastics
Catalog # 97002-256
Flexible Vent Assemblies, Scientific Plastics
Double cabinet penetration, Kewaunee* cabinets with Labconco* worsurfaces
Price
$526.22
Log in pour la disponibilité
Each
Product Details & Documents
Venting systems are designed for use in acid and solvent cabinets.
  • All vents are manufactured from acid-resistant polyolefin material
  • Non-squeezed top
The flexible tubing design allows systems to be installed around potential obstacles within existing cabinet structures. A 3 m (10') length of accessory tubing is available separately for field modifications.
Spécifications
  • Désignation:
    Double cabinet penetration
  • For Use With:
    Kewaunee* cabinets with Labconco* worsurfaces
Accessories
Informations sur le produit
  • Catalog #
    Length
    Availability
    Price
  • Flexible pipe
    Flexible pipe
    97002-258
    10 ft.
    Log in pour la disponibilité
    Each - $397.49