Flexible Vent Assemblies, Scientific Plastics
Double cabinet penetration, Kewaunee* cabinets with Labconco* worsurfaces
Catalog # 97002-256
Supplier: Scientific Plastics
: F160L078F-150
Jump to:
Product Details & Documents
Venting systems are designed for use in acid and solvent cabinets.
- All vents are manufactured from acid-resistant polyolefin material
- Non-squeezed top
The flexible tubing design allows systems to be installed around potential obstacles within existing cabinet structures. A 3 m (10') length of accessory tubing is available separately for field modifications.
Spécifications
- Désignation:Double cabinet penetration
- For Use With:Kewaunee* cabinets with Labconco* worsurfaces
Accessories
Informations sur le produit
- Catalog #LengthAvailabilityPrice
- Flexible pipe