Control Slides, Azer Scientific | Avantor
Control Slides, Azer Scientific
Control slides, Melanin, 75×25 mm (3×1")
Catalog # 95042-318
Fournisseur #:  ESCS-122
Control Slides, Azer Scientific
Catalog # 95042-318
Control Slides, Azer Scientific
Control Slides, Azer Scientific
Control slides, Melanin, 75×25 mm (3×1")
Price
$164.19
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 10
Product Details & Documents
Control slides are designed for use in monitoring stain performance in the Pathology lab.
  • Permanent positive charge for improved readability
  • Control slides with paraffin-embedded human tissue sections
  • Ten unstained Unifrost slides
  • One stained slide
All slides feature a permanent positive charge that securely anchors all types of sections and improves the quality of preparations and readability
Includes 10 unstained Unifrost microscope slides and one additional slide, which is stained with the proper procedure to demonstrate and validate the expected result.
Azer Scientific's control slides are carefully prepared with paraffin-embedded human tissue sections.
 : 11 per pack
Spécifications
  • Désignation:
    Control slides, Melanin
  • L×l:
    75×25 mm (3×1")