NMC Valve Wheel Lockouts
Fits 25.4 cm (10") To 35.56 cm (14") Dia.
Catalog # 94022-912
Supplier: ACCUFORM MANUFACTURING, INC
Fournisseur #: VL13R
Jump to:
Product Details & Documents
Lockouts consist of two flattened half moons that completely cover the valve wheels from 1" to 14" in diameter
Made from strong Polypro plastic, fully dielectric. Highly resistant to cracking and abrasion. Resistant to extreme temperature changes 50⁰F
Spécifications
- Désignation:Fits 25.4 cm (10") To 35.56 cm (14") Dia.