NMC Valve Wheel Lockouts
Fits 16.51 cm (6.5") To 25.4 cm (10") Dia.
Catalog # 94022-910
Supplier: ACCUFORM MANUFACTURING, INC
Supplier #: VL09R
Jump to:
Product Details & Documents
Lockouts consist of two flattened half moons that completely cover the valve wheels from 1" to 14" in diameter
Made from strong Polypro plastic, fully dielectric. Highly resistant to cracking and abrasion. Resistant to extreme temperature changes 50⁰F
Specifications
- Désignation:Fits 16.51 cm (6.5") To 25.4 cm (10") Dia.