Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
MasterLock® Lockout Padlocks, NMC

MasterLock® Lockout Padlocks, NMC

1.91 cm (3/4"), Green
Catalog # 94017-584
Supplier #:  MPSG
MasterLock® Lockout Padlocks, NMC
Catalog # 94017-584
MasterLock® Lockout Padlocks, NMC
MasterLock® Lockout Padlocks, NMC

1.91 cm (3/4"), Green

Price
$88.93
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Over 10,000 unique key codes, 4 pin tumbler cylinders

Heavy duty laminated steel case constructed with hardened shackles. Full keying capability for most commercial applications. Locks are keyed differently. Two keys included.

Specifications
  • Color:
    Green
  • Shackle Size:
    1.91 cm (3/4")