Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
MasterLock® Lockout Padlocks, NMC | Avantor

MasterLock® Lockout Padlocks, NMC

3.81 cm (11/2"), White
Catalog # 94017-576
Fournisseur #:  MPLW
MasterLock® Lockout Padlocks, NMC
Catalog # 94017-576
MasterLock® Lockout Padlocks, NMC
MasterLock® Lockout Padlocks, NMC

3.81 cm (11/2"), White

Price
$102.90
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents

Over 10,000 unique key codes, 4 pin tumbler cylinders

Heavy duty laminated steel case constructed with hardened shackles. Full keying capability for most commercial applications. Locks are keyed differently. Two keys included.

Spécifications
  • Color:
    White
  • Shackle Size:
    3.81 cm (11/2")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.