MasterLock® Lockout Padlocks, NMC
3.81 cm (11/2"), Green
94017-572
: MPLG
Over 10,000 unique key codes, 4 pin tumbler cylinders
Heavy duty laminated steel case constructed with hardened shackles. Full keying capability for most commercial applications. Locks are keyed differently. Two keys included.
- Color:Green
- Shackle Size:3.81 cm (11/2")