NMC Electrical Lockout Center
Electrical Lockout Backboard And Rack
Catalog # 94016-422
Jump to:
Discontinued
Product Details & Documents
Made from .050 rigid styrene with four corner mounting holes
Wire basket holds 12" toolbox and components 40.64cm x 35.56cm (16" x 14"). Available with or without contents.
Spécifications
- Désignation:Electrical Lockout Backboard And Rack