Utility Posts, National Marker | Avantor
Utility Posts, National Marker
122 cm (4'), Blue
Catalog # 94013-728
Fournisseur #:  EZ4BL
Utility Posts, National Marker
Catalog # 94013-728
Utility Posts, National Marker
Utility Posts, National Marker
122 cm (4'), Blue
Price
$213.60
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
Protect your valuable under-ground investment
The only post that can be tied in a knot, yet is so stiff it can withstand high wind loading. Well suited for marking pipelines, conduits and cables. Standard height is 4' (above ground), standard width 8.26cm (3.25"). Available in orange, green, yellow, blue, red and brown.
Spécifications
  • Color:
    Blue
  • Dimensions:
    122 cm (4')