NMC Ball Valve Lockouts | Avantor
NMC Ball Valve Lockouts
Fits 0.95 cm (3/8") To 3.18 cm (11/4")
Catalog # 94004-342
Fournisseur #:  BS01R
NMC Ball Valve Lockouts
Catalog # 94004-342
NMC Ball Valve Lockouts
NMC Ball Valve Lockouts
Fits 0.95 cm (3/8") To 3.18 cm (11/4")
Price
190,39 $
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
Lock ball valves safety, efficiently and inexpensively
Available in 3 sizes. Fits .312" diameter hole.
Spécifications
  • Taille:
    Fits 0.95 cm (3/8") To 3.18 cm (11/4")