Dissolution Calibration Tools, Quality Lab Accessories
1 cm Stationary Basket Spacer for Legacy VanKel Baths
Catalog # 93001-072
Supplier: QLA
Fournisseur #: DEPSET-SB
Jump to:
Product Details & Documents
QLA manufactures Calibrations tools that have been designed for use on most dissolution baths.
- Mechanical calibration parameters are easily verified using QLA's family of Calibration Tools
All are supplied with NIST traceable calibration certificates and user manuals. These calibration tools are serialized and come with a 1 year calibration certificate. Recalibration services are also offered by QLA.
Certifications: All calibration tools are serialized and come with a 1 year NIST traceable calibration certificate.
Spécifications
- Désignation:1 cm Stationary Basket Spacer for Legacy VanKel Baths