Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Corning® Solid Black and White Polystyrene Microplates, 384 Well

Corning® Solid Black and White Polystyrene Microplates, 384 Well

Solid plates with lid and bar code, 112 µl, 112 µl, 20 - 80 µl, White, Sterile, Tissue culture (TC), Flat-Bottom, 10/Bag
Catalog # 89176-442
Product available on GSA Advantage®
   Low Manufacturing Impact
Supplier:   Corning
Supplier #:  3570BC
Corning® Solid Black and White Polystyrene Microplates, 384 Well
Catalog # 89176-442
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
Corning® Solid Black and White Polystyrene Microplates, 384 Well
Corning® Solid Black and White Polystyrene Microplates, 384 Well

Solid plates with lid and bar code, 112 µl, 112 µl, 20 - 80 µl, White, Sterile, Tissue culture (TC), Flat-Bottom, 10/Bag

Price
$733.55
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 50
Product Details & Documents

PS. Designed to reduce well-to-well crosstalk during fluorescent and luminescent assays.

  • Square well top for easier robotic pipetting
  • Round bottom for better Z factor and minimized trapped air
  • Conical well molded in the shape of a light cone for efficiency
  • Black walls reduce well-to-well crosstalk and autofluorescence during fluorescent assays

High binding surface is a general purpose surface that binds medium (>10kD) and large biomolecules possessing ionic groups and/or hydrophobic regions. Binding capacity is approximately 400 to 500ng IgG/cm.

Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions. Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface (binding capacity of approximately 100 to 200ng IgG/cm²).

Nonbinding surface (NBS™) is a Corning proprietary treatment technology used on polystyrene microplates to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions. Ideal for reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise.

Spécifications
  • Sterility:
    Sterile
  • Well Volume:
    112 µl
  • Color:
    White
  • Désignation:
    Solid plates with lid and bar code
  • Packaging:
    10/Bag
  • Well Shape:
    Flat-Bottom
  • Surface Treatment:
    Tissue culture (TC)
  • Working Well Volume:
    20 - 80 µl
  • Total Well Volume:
    112 µl
  • Cat. No.:
    89176-442
  • Environmental certification:
    RECs, ISO 14001 - Kennebunk
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.