Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Protector® Filtered Glove Box, Labconco | Avantor

Protector® Filtered Glove Box, Labconco

ULPA Filtered, Single Width, 208/230 V, 50/60 Hz (China/Australia), Stainless steel
Catalog # 89175-298
Supplier:   Labconco
Fournisseur #:  5065540
Protector® Filtered Glove Box, Labconco
Catalog # 89175-298
Supplier:  Labconco
Protector® Filtered Glove Box, Labconco
Protector® Filtered Glove Box, Labconco

ULPA Filtered, Single Width, 208/230 V, 50/60 Hz (China/Australia), Stainless steel

Price
$39,520.00
Log in pour la disponibilité
Each
Product Details & Documents

Protector® Filtered Glove Boxes provide inlet and outlet HEPA or ULPA filtration and a leak-tight physical barrier to protect the operator from exposure to potentially hazardous materials

These glove boxes feature particulate containment <20 nanograms per cubic meter as confirmed by Safebridge Consultants, Incorporated and also meet ISO Class 3 standards.

Informations sur les accessoires: Glove box interior shelves kit provides shelf space across the back wall. Three stainless steel storage shelves are attached to epoxy-coated steel upright supports. Shelves have adjustable height. Installation hardware and instructions are included.

Spécifications
  • Désignation:
    ULPA Filtered, Single Width
  • Matière:
    Stainless steel
  • Electrical:
    208/230 V, 50/60 Hz (China/Australia)
  • Certification:
    CE
Accessories

Informations sur le produit

  • Catalog #
    Availability
    Price
  • Glove Box Interior Shelves Kit, 76.2W x 15.2D cm (30 x 6")
    Glove Box Interior Shelves Kit, 76.2W x 15.2D cm (30 x 6")
    32940-080
    Log in pour la disponibilité
    Each - $1,120.00

Informations sur le produit

  • Catalog #
    Availability
    Price
  • Add-A-Valve Kit, Two Manual Valves for Transfer Chamber
    Add-A-Valve Kit, Two Manual Valves for Transfer Chamber
    89194-714
    Log in pour la disponibilité
    Each - $2,420.00

Informations sur le produit

  • Catalog #
    Availability
    Price
  • Add-A-Valve Kit, Two Manual Valves for Main Chamber
    Add-A-Valve Kit, Two Manual Valves for Main Chamber
    89194-716
    Log in pour la disponibilité
    Each - $2,060.00

Informations sur le produit

  • Catalog #
    Availability
    Price
  • Recirculation Kit
    Recirculation Kit
    89194-712
    Log in pour la disponibilité
    Each - $1,050.00

Informations sur le produit

  • Catalog #
    Availability
    Price
  • Hard Duct Connector
    Hard Duct Connector
    89194-710
    Log in pour la disponibilité
    Each - $896.00

Informations sur le produit

  • Catalog #
    Availability
    Price
  • Thimble Exhaust Connector, 6", (For connection to house exhaust or remote blower)
    Thimble Exhaust Connector, 6", (For connection to house exhaust or remote blower)
    89194-708
    Log in pour la disponibilité
    Each - $1,130.00

Informations sur le produit

  • Catalog #
    Availability
    Price
  • Interior Glove Port Cover
    Interior Glove Port Cover
    15202-521
    Log in pour la disponibilité
    Each - $2,380.00

Informations sur le produit

  • Catalog #
    Availability
    Price
  • HEPA Inlet/Exhaust Filter
    HEPA Inlet/Exhaust Filter
    32940-085
    Log in pour la disponibilité
    Each - $1,130.00

Informations sur le produit

  • Catalog #
    Availability
    Price
  • Carbon Exhaust Filter
    Carbon Exhaust Filter
    32940-090
    Log in pour la disponibilité
    Each - $2,060.00

Informations sur le produit

  • Catalog #
    Availability
    Price
  • Exterior Monitor Tubing Kit (Use with Moisture Monitors 5244900 and 5244901)
    Exterior Monitor Tubing Kit (Use with Moisture Monitors 5244900 and 5244901)
    89175-442
    Log in pour la disponibilité
    Each - $427.00

Informations sur le produit

  • Catalog #
    Availability
    Price
  • Add-A-Valve Installation Kit
    Add-A-Valve Installation Kit
    89194-724
    Log in pour la disponibilité
    Each - $161.00

Informations sur le produit

  • Catalog #
    Availability
    Price
  • Blower Foot Switch
    Blower Foot Switch
    89194-706
    Log in pour la disponibilité
    Each - $1,050.00

Informations sur le produit

  • Catalog #
    Availability
    Price
  • Positive Pressure Conversion Kit
    Positive Pressure Conversion Kit
    89194-704
    Log in pour la disponibilité
    Each - $1,050.00

Informations sur le produit

  • Catalog #
    Availability
    Price
  • Thimble Exhaust Connectors, 5" (For conection to FilterMate Portable Exhauster)
    Thimble Exhaust Connectors, 5" (For conection to FilterMate Portable Exhauster)
    89200-778
    Log in pour la disponibilité
    Each - $1,440.00