Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Protector® Filtered Glove Box, Labconco

Protector® Filtered Glove Box, Labconco

ULPA Filtered, Single Width, 115 V, 50/60 Hz (US), Stainless steel
Catalog # 89175-292
Supplier:   Labconco
Supplier #:  5065510
Protector® Filtered Glove Box, Labconco
Catalog # 89175-292
Supplier:  Labconco
Protector® Filtered Glove Box, Labconco
Protector® Filtered Glove Box, Labconco

ULPA Filtered, Single Width, 115 V, 50/60 Hz (US), Stainless steel

Price
$39,520.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Protector® Filtered Glove Boxes provide inlet and outlet HEPA or ULPA filtration and a leak-tight physical barrier to protect the operator from exposure to potentially hazardous materials

These glove boxes feature particulate containment <20 nanograms per cubic meter as confirmed by Safebridge Consultants, Incorporated and also meet ISO Class 3 standards.

Accessories information: Glove box interior shelves kit provides shelf space across the back wall. Three stainless steel storage shelves are attached to epoxy-coated steel upright supports. Shelves have adjustable height. Installation hardware and instructions are included.

Specifications
  • Désignation:
    ULPA Filtered, Single Width
  • Matière:
    Stainless steel
  • Electrical:
    115 V, 50/60 Hz (US)
  • Certification:
    UL, ETL, CAN/CSA
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Glove Box Interior Shelves Kit, 76.2W x 15.2D cm (30 x 6")
    Glove Box Interior Shelves Kit, 76.2W x 15.2D cm (30 x 6")
    32940-080
    Log in for availability
    Each - $1,120.00

Product Information

  • Catalog #
    Availability
    Price
  • Add-A-Valve Kit, Two Manual Valves for Transfer Chamber
    Add-A-Valve Kit, Two Manual Valves for Transfer Chamber
    89194-714
    Log in for availability
    Each - $2,420.00

Product Information

  • Catalog #
    Availability
    Price
  • Add-A-Valve Kit, Two Manual Valves for Main Chamber
    Add-A-Valve Kit, Two Manual Valves for Main Chamber
    89194-716
    Log in for availability
    Each - $2,060.00

Product Information

  • Catalog #
    Availability
    Price
  • Recirculation Kit
    Recirculation Kit
    89194-712
    Log in for availability
    Each - $1,050.00

Product Information

  • Catalog #
    Availability
    Price
  • Hard Duct Connector
    Hard Duct Connector
    89194-710
    Log in for availability
    Each - $896.00

Product Information

  • Catalog #
    Availability
    Price
  • Thimble Exhaust Connector, 6", (For connection to house exhaust or remote blower)
    Thimble Exhaust Connector, 6", (For connection to house exhaust or remote blower)
    89194-708
    Log in for availability
    Each - $1,130.00

Product Information

  • Catalog #
    Availability
    Price
  • Interior Glove Port Cover
    Interior Glove Port Cover
    15202-521
    Log in for availability
    Each - $2,380.00

Product Information

  • Catalog #
    Availability
    Price
  • HEPA Inlet/Exhaust Filter
    HEPA Inlet/Exhaust Filter
    32940-085
    Log in for availability
    Each - $1,130.00

Product Information

  • Catalog #
    Availability
    Price
  • Carbon Exhaust Filter
    Carbon Exhaust Filter
    32940-090
    Log in for availability
    Each - $2,060.00

Product Information

  • Catalog #
    Availability
    Price
  • Exterior Monitor Tubing Kit (Use with Moisture Monitors 5244900 and 5244901)
    Exterior Monitor Tubing Kit (Use with Moisture Monitors 5244900 and 5244901)
    89175-442
    Log in for availability
    Each - $427.00

Product Information

  • Catalog #
    Availability
    Price
  • Add-A-Valve Installation Kit
    Add-A-Valve Installation Kit
    89194-724
    Log in for availability
    Each - $161.00

Product Information

  • Catalog #
    Availability
    Price
  • Blower Foot Switch
    Blower Foot Switch
    89194-706
    Log in for availability
    Each - $1,050.00

Product Information

  • Catalog #
    Availability
    Price
  • Positive Pressure Conversion Kit
    Positive Pressure Conversion Kit
    89194-704
    Log in for availability
    Each - $1,050.00

Product Information

  • Catalog #
    Availability
    Price
  • Thimble Exhaust Connectors, 5" (For conection to FilterMate Portable Exhauster)
    Thimble Exhaust Connectors, 5" (For conection to FilterMate Portable Exhauster)
    89200-778
    Log in for availability
    Each - $1,440.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.