Protector® Filtered Glove Box, Labconco
HEPA Filtered, Single Width, 115 V, 50/60 Hz (US), Fiberglass
89175-282
: Labconco
: 5065010
Protector® Filtered Glove Boxes provide inlet and outlet HEPA or ULPA filtration and a leak-tight physical barrier to protect the operator from exposure to potentially hazardous materials
These glove boxes feature particulate containment <20 nanograms per cubic meter as confirmed by Safebridge Consultants, Incorporated and also meet ISO Class 3 standards.
: Glove box interior shelves kit provides shelf space across the back wall. Three stainless steel storage shelves are attached to epoxy-coated steel upright supports. Shelves have adjustable height. Installation hardware and instructions are included.
- Désignation:HEPA Filtered, Single Width
- Matière:Fiberglass
- Electrical:115 V, 50/60 Hz (US)
- Certification:UL, ETL, CAN/CSA
Accessories
- Description
- Description
- Description
- Description
- Description
- Description
- 89194-708Thimble Exhaust Connector, 6", (For connection to house exhaust or remote blower)Each - $1,130.00
- Description
- Description
- Description
- Description
- 89175-442Exterior Monitor Tubing Kit (Use with Moisture Monitors 5244900 and 5244901)Each - $427.00
- Description
- Description
- Description
- Description
- 89200-778Thimble Exhaust Connectors, 5" (For conection to FilterMate Portable Exhauster)Each - $1,440.00



