Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
General Printer Labels, Brady

General Printer Labels, Brady

Printer labels, B-569, Polyester, Yellow, BBP®31, BBP®33, 1.125", 30.48 m (100 ft.)
Catalog # 89174-930
Supplier:   Brady Worldwide
Supplier #:  117992
General Printer Labels, Brady
Catalog # 89174-930
Supplier:  Brady Worldwide
General Printer Labels, Brady
General Printer Labels, Brady

Printer labels, B-569, Polyester, Yellow, BBP®31, BBP®33, 1.125", 30.48 m (100 ft.)

Price
$508.09
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Roll of 1
Product Details & Documents

General labels for straws, petri dishes, bottles, flasks, microplates, bags, and more.

Tamper evident labels and tapes: B-438 metalized polyester, permanent adhesive designed to leave checkboard footprint if removed.

Spécifications
  • Pour l'utilisation avec:
    BBP®31, BBP®33
  • Color:
    Yellow
  • Désignation:
    Printer labels, B-569
  • Matière:
    Polyester
  • Roll Length:
    30.48 m (100 ft.)
  • Width:
    1.125"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.