Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

General Printer Labels, Brady

Printer labels, B-569, Polyester, Blue, BBP®31, BBP®33, 0.5", 30.48 m (100 ft.)
  89174-878
 :  117936
General Printer Labels, Brady
  89174-878
General Printer Labels, Brady
General Printer Labels, Brady

Printer labels, B-569, Polyester, Blue, BBP®31, BBP®33, 0.5", 30.48 m (100 ft.)

 
$338.37
 
   
Roll of 1
 

General labels for straws, petri dishes, bottles, flasks, microplates, bags, and more.

Tamper evident labels and tapes: B-438 metalized polyester, permanent adhesive designed to leave checkboard footprint if removed.

 
  • Pour l'utilisation avec:
    BBP®31, BBP®33
  • Color:
    Blue
  • Désignation:
    Printer labels, B-569
  • Matière:
    Polyester
  • Roll Length:
    30.48 m (100 ft.)
  • Width:
    0.5"