Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

General Printer Labels, Brady

Printer labels, B-526, Phosphorescent Polyester, Photoluminescent, BBP®31, BBP®33, 4", 15.24 m (50 ft.)
Catalog # 89174-844
Supplier:   Brady Worldwide
 :  117924
General Printer Labels, Brady
Catalog # 89174-844
Supplier:  Brady Worldwide
General Printer Labels, Brady
General Printer Labels, Brady

Printer labels, B-526, Phosphorescent Polyester, Photoluminescent, BBP®31, BBP®33, 4", 15.24 m (50 ft.)

Price
$1,483.87
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Roll of 1
Product Details & Documents

General labels for straws, petri dishes, bottles, flasks, microplates, bags, and more.

Tamper evident labels and tapes: B-438 metalized polyester, permanent adhesive designed to leave checkboard footprint if removed.

Spécifications
  • Pour l'utilisation avec:
    BBP®31, BBP®33
  • Color:
    Photoluminescent
  • Désignation:
    Printer labels, B-526
  • Matière:
    Phosphorescent Polyester
  • Roll Length:
    15.24 m (50 ft.)
  • Width:
    4"