Explorer® Precision Balances, Ohaus®
4200 g, ±0.02 g, 190×200 mm (71/2 × 77/8"), 0.01 g, 1 s, 23×9.8×39.3 cm
Catalog # 89171-462
 :  83021357
Explorer® Precision Balances, Ohaus®
Catalog # 89171-462
Supplier:  
Explorer® Precision Balances, Ohaus®
Explorer® Precision Balances, Ohaus®
4200 g, ±0.02 g, 190×200 mm (71/2 × 77/8"), 0.01 g, 1 s, 23×9.8×39.3 cm
Discontinued
Product Details & Documents
Precision balances provide accurate results in seconds to improve efficiency and productivity
Four touchless sensors allow for hands-free operation of print, tare, calibration, and other functions. All balances are equipped with AutoCal™ automatic internal calibration adjustment system, which self-calibrates the system when it senses a temperature change significant enough to affect weighing accuracy or every 11 hours. SmarText™ 2.0 graphical software features 14 applications, QWERTY and numeric keypads, and below-minimum weight indication. Large color touch screen displays allow for easy menu navigation. The display includes USB and RS-232 ports, and can be separated from the weighing base for maximum flexibility in a broad range of applications. The weighing base features a QuadraStance™ design with four adjustable thumbwheels to provide superior stability. All models offering a readability of 0.001g feature a frameless, flip-top draftshield constructed of antistatic coated glass
 : CE marked. FCC, CMC, OIML Class I and Class II Approved.
 : Balances are supplied with two-year manufacturer's warranty, AC adapter, weighing pan, pan support, multilanguage operator's manual with quick-start guide, and in-use cover.
Spécifications
  • Plateau de pesée/taille de la plateforme:
    190×200 mm (71/2 × 77/8")
  • Dimensions:
    23×9.8×39.3 cm
  • Linearity:
    ±0.02 g
  • Stabilization Time:
    1 s
  • Readability:
    0.01 g
  • Weighing Capacity:
    4200 g
  • Cat. No.:
    89171-462
  • Voltage:
    100–240V, 50–60Hz
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Anti-Theft Device (Laptop lock), for Explorer®/Explorer® Pro/Triple Beam/MB Series
    Anti-Theft Device (Laptop lock), for Explorer®/Explorer® Pro/Triple Beam/MB Series
    11380-322
    Log in pour la disponibilité
    Each - $567.24
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Printer Replacement Cable, for all Explorer® Series
    Printer Replacement Cable, for all Explorer® Series
    11380-304
    Log in pour la disponibilité
    Each - $212.10
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Ethernet Interface Kit, for all Explorer® Series
    Ethernet Interface Kit, for all Explorer® Series
    89194-604
    Log in pour la disponibilité
    Each - $473.01
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Cable, USB Interface, (Type A to B), for Explorer®/Adventurer® Series
    Cable, USB Interface, (Type A to B), for Explorer®/Adventurer® Series
    89194-600
    Log in pour la disponibilité
    Each - $473.01
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Printer Replacement Ink Ribbon, for all Explorer® Series
    Printer Replacement Ink Ribbon, for all Explorer® Series
    89496-992
    Log in pour la disponibilité
    Each - $499.43
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Rechargeable Battery, EX-HiCap R71, for all Explorer® Series
    Rechargeable Battery, EX-HiCap R71, for all Explorer® Series
    89496-996
    Log in pour la disponibilité
    Each - $1,346.39
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Tower Mount for Terminal, for all Explorer® Series
    Tower Mount for Terminal, for all Explorer® Series
    89194-592
    Log in pour la disponibilité
    Each - $408.64
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Terminal Extension Cable, for all Explorer® Series
    Terminal Extension Cable, for all Explorer® Series
    89194-594
    Log in pour la disponibilité
    Each - $266.00
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Wheel Kit for EX-HiCap (Set of 4), for all Explorer® Series
    Wheel Kit for EX-HiCap (Set of 4), for all Explorer® Series
    89496-998
    Log in pour la disponibilité
    Each - $411.34
Informations sur le produit
  • Catalog #
    Availability
    Price
  • RS232 Cable, PC 25 Pin, for Discovery/Explorer®/Adventurer™ Pro/Pioneer™ Series
    RS232 Cable, PC 25 Pin, for Discovery/Explorer®/Adventurer™ Pro/Pioneer™ Series
    89194-608
    Log in pour la disponibilité
    Each - $473.01
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Paper Roll for Printer SF40A, 57.5 mm Width, for Explorer®/Explorer® Pro/Discovery/Adventurer™ Pro/Pioneer™/Defender™/CKW/Ranger™/Valor™/Navigator®/Scout® Pro/MB Series
    Paper Roll for Printer SF40A, 57.5 mm Width, for Explorer®/Explorer® Pro/Discovery/Adventurer™ Pro/Pioneer™/Defender™/CKW/Ranger™/Valor™/Navigator®/Scout® Pro/MB Series
    89497-004
    Log in pour la disponibilité
    Each - $65.00
Informations sur le produit
  • Catalog #
    Availability
    Price
  • RS232 Cable & Adapter to STP103 Printer, for Discovery/Explorer®/Adventurer™ Pro/Pioneer™/Defender™ 3000/5000/MB Series
    RS232 Cable & Adapter to STP103 Printer, for Discovery/Explorer®/Adventurer™ Pro/Pioneer™/Defender™ 3000/5000/MB Series
    97009-696
    Log in pour la disponibilité
    Each - $518.49
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Density Kit, Solids, for all Explorer® Series
    Density Kit, Solids, for all Explorer® Series
    89194-596
    Log in pour la disponibilité
    Each - $1,896.26