Explorer® Precision Balances, Ohaus®
Product 76097-972 is similar to 89171-460 but manufacturer and specifications may vary.
Product 76097-974 is similar to 89171-460 but manufacturer and specifications may vary.
Product Details & Documents
Precision balances provide accurate results in seconds to improve efficiency and productivity
Four touchless sensors allow for hands-free operation of print, tare, calibration, and other functions. All balances are equipped with AutoCal™ automatic internal calibration adjustment system, which self-calibrates the system when it senses a temperature change significant enough to affect weighing accuracy or every 11 hours. SmarText™ 2.0 graphical software features 14 applications, QWERTY and numeric keypads, and below-minimum weight indication. Large color touch screen displays allow for easy menu navigation. The display includes USB and RS-232 ports, and can be separated from the weighing base for maximum flexibility in a broad range of applications. The weighing base features a QuadraStance™ design with four adjustable thumbwheels to provide superior stability. All models offering a readability of 0.001g feature a frameless, flip-top draftshield constructed of antistatic coated glass
Certifications: CE marked. FCC, CMC, OIML Class I and Class II Approved.
Ordering information: Balances are supplied with two-year manufacturer's warranty, AC adapter, weighing pan, pan support, multilanguage operator's manual with quick-start guide, and in-use cover.
Spécifications
- Plateau de pesée/taille de la plateforme:190×200 mm (71/2 × 77/8")
- Dimensions:23×9.8×39.3 cm
- Linearity:±0.02 g
- Stabilization Time:1 s
- Readability:0.01 g
- Weighing Capacity:2000 g
- Cat. No.:89171-460
- Voltage:100–240V, 50–60Hz
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Anti-Theft Device (Laptop lock), for Explorer®/Explorer® Pro/Triple Beam/MB SeriesAnti-Theft Device (Laptop lock), for Explorer®/Explorer® Pro/Triple Beam/MB Series11380-322Log in pour la disponibilitéEach - $567.24
Informations sur le produit
- Catalog #AvailabilityPrice
- Printer Replacement Cable, for all Explorer® SeriesPrinter Replacement Cable, for all Explorer® Series11380-304Log in pour la disponibilitéEach - $212.10
Informations sur le produit
- Catalog #AvailabilityPrice
- Ethernet Interface Kit, for all Explorer® SeriesEthernet Interface Kit, for all Explorer® Series89194-604Log in pour la disponibilitéEach - $473.01
Informations sur le produit
- Catalog #AvailabilityPrice
- Cable, USB Interface, (Type A to B), for Explorer®/Adventurer® SeriesCable, USB Interface, (Type A to B), for Explorer®/Adventurer® Series89194-600Log in pour la disponibilitéEach - $473.01
Informations sur le produit
- Catalog #AvailabilityPrice
- Printer Replacement Ink Ribbon, for all Explorer® SeriesPrinter Replacement Ink Ribbon, for all Explorer® Series89496-992Log in pour la disponibilitéEach - $499.43
Informations sur le produit
- Catalog #AvailabilityPrice
- Rechargeable Battery, EX-HiCap R71, for all Explorer® SeriesRechargeable Battery, EX-HiCap R71, for all Explorer® Series89496-996Log in pour la disponibilitéEach - $1,346.39
Informations sur le produit
- Catalog #AvailabilityPrice
- Tower Mount for Terminal, for all Explorer® SeriesTower Mount for Terminal, for all Explorer® Series89194-592Log in pour la disponibilitéEach - $408.64
Informations sur le produit
- Catalog #AvailabilityPrice
- Terminal Extension Cable, for all Explorer® SeriesTerminal Extension Cable, for all Explorer® Series89194-594Log in pour la disponibilitéEach - $266.00
Informations sur le produit
- Catalog #AvailabilityPrice
- Wheel Kit for EX-HiCap (Set of 4), for all Explorer® SeriesWheel Kit for EX-HiCap (Set of 4), for all Explorer® Series89496-998Log in pour la disponibilitéEach - $411.34
Informations sur le produit
- Catalog #AvailabilityPrice
- RS232 Cable, PC 25 Pin, for Discovery/Explorer®/Adventurer™ Pro/Pioneer™ SeriesRS232 Cable, PC 25 Pin, for Discovery/Explorer®/Adventurer™ Pro/Pioneer™ Series89194-608Log in pour la disponibilitéEach - $473.01
Informations sur le produit
- Catalog #AvailabilityPrice
- Paper Roll for Printer SF40A, 57.5 mm Width, for Explorer®/Explorer® Pro/Discovery/Adventurer™ Pro/Pioneer™/Defender™/CKW/Ranger™/Valor™/Navigator®/Scout® Pro/MB SeriesPaper Roll for Printer SF40A, 57.5 mm Width, for Explorer®/Explorer® Pro/Discovery/Adventurer™ Pro/Pioneer™/Defender™/CKW/Ranger™/Valor™/Navigator®/Scout® Pro/MB Series89497-004Log in pour la disponibilitéEach - $65.00
Informations sur le produit
- Catalog #AvailabilityPrice
- RS232 Cable & Adapter to STP103 Printer, for Discovery/Explorer®/Adventurer™ Pro/Pioneer™/Defender™ 3000/5000/MB SeriesRS232 Cable & Adapter to STP103 Printer, for Discovery/Explorer®/Adventurer™ Pro/Pioneer™/Defender™ 3000/5000/MB Series97009-696Log in pour la disponibilitéEach - $518.49
Informations sur le produit
- Catalog #AvailabilityPrice
- Density Kit, Solids, for all Explorer® SeriesDensity Kit, Solids, for all Explorer® Series89194-596Log in pour la disponibilitéEach - $1,896.26