Duramark® Lock Boxes
Plastic lock box, 34.3×26×17.1 cm (13½×10¼× 6¾”)
89170-916
: 36745
Metal lock boxes are constructed of 24 gauge cold-rolled steel, painted with a powder coat finish, and insulated with foam
Plastic lock boxes are made of high impact polystrene and feature a viewing window on the door. Both boxes feature a hinged door that opens downward for convenient access and use as a tray. Two matching keys are supplied. The metal lock box is also available with a combination lock. Over- or under-door straps allow the boxes to be secured outside of a locked door for easy courier access.
- Désignation:Plastic lock box
- Dimensions:34.3×26×17.1 cm (13½×10¼× 6¾”)
Accessories
- Description
- Description
- Description
- Description