Duramark® Lock Boxes
Metal lock box with under-door strap, 35.5×26.7×17.8 cm (14×10½×7”)
Catalog # 89170-914
Supplier: Therapak, LLC
Fournisseur #: 36744
Jump to:
Product Details & Documents
Metal lock boxes are constructed of 24 gauge cold-rolled steel, painted with a powder coat finish, and insulated with foam
Plastic lock boxes are made of high impact polystrene and feature a viewing window on the door. Both boxes feature a hinged door that opens downward for convenient access and use as a tray. Two matching keys are supplied. The metal lock box is also available with a combination lock. Over- or under-door straps allow the boxes to be secured outside of a locked door for easy courier access.
Spécifications
- Désignation:Metal lock box with under-door strap
- Dimensions:35.5×26.7×17.8 cm (14×10½×7”)
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Under-Door Strap for Metal or Plastic Box
Informations sur le produit
- Catalog #AvailabilityPrice
- Over-Door Strap for Metal or Plastic Box
Informations sur le produit
- Catalog #AvailabilityPrice
- Extra Long Over-Door Strap for Metal or Plastic BoxExtra Long Over-Door Strap for Metal or Plastic Box10497-106Log in pour la disponibilitéEach - $188.06
Informations sur le produit
- Catalog #AvailabilityPrice
- Replacement Key for Metal or Plastic Box
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



