Corning® Bar Coded Storage Tubes, Corning
384× 2D barcoded storage tubes, square, without plug caps, Black, 120 µl, racked
Catalog # 89168-526
Supplier: Corning
Fournisseur #: 8510
Jump to:
Product Details & Documents
PP. The 1D/2D storage line is an innovative product with both a linear 1D and 2D barcodes etched on each tube.
- Maximum information, 14×14 dot 2D barcode
- Barcode stability, laser-etched, not an attached label
- Compound compatibility, polypropylene construction throughout, making it inert
Spécifications
- Volume:120 µl
- Désignation:384× 2D barcoded storage tubes, square, without plug caps, Black
- Packaging:racked
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- 96 screw caps for storage tubes 1,3 ml, with O-Ring, Orange, for no. 850096 screw caps for storage tubes 1,3 ml, with O-Ring, Orange, for no. 850089168-514Log in pour la disponibilitéCase of 4,800 - $5,060.00
Informations sur le produit
- Catalog #AvailabilityPrice
- 96 storage tube screw cap, with O-Ring, on mat, clear with cover, Orange, for no. 850096 storage tube screw cap, with O-Ring, on mat, clear with cover, Orange, for no. 850089168-516Log in pour la disponibilitéCase of 4,800 - $5,880.00
Informations sur le produit
- Catalog #AvailabilityPrice
- Thermoplastic elastomer (TPE) 96 storage tube septum cap, White, for no. 8500Thermoplastic elastomer (TPE) 96 storage tube septum cap, White, for no. 850089168-518Log in pour la disponibilitéCase of 9,600 - $901.00
Informations sur le produit
- Catalog #AvailabilityPrice
- Thermoplastic elastomer (TPE) 96 storage tube septum cap on mat, White, for no. 8500Thermoplastic elastomer (TPE) 96 storage tube septum cap on mat, White, for no. 850089168-520Log in pour la disponibilitéCase of 4,800 - $1,480.00
Informations sur le produit
- Catalog #AvailabilityPrice
- Empty racks with lids for 96 screw capped storage tubes no. 8502, NaturalEmpty racks with lids for 96 screw capped storage tubes no. 8502, Natural89168-522Log in pour la disponibilitéCase of 10 - $145.00
Informations sur le produit
- Catalog #AvailabilityPrice
- 384 storage tube plug cap on mats for round tube no. 8509, 384 caps per mat, Orange384 storage tube plug cap on mats for round tube no. 8509, 384 caps per mat, Orange89168-530Log in pour la disponibilitéCase of 38,400 - $7,100.00
Informations sur le produit
- Catalog #AvailabilityPrice
- 384 Storage Tube Plug Cap,Orange, for no. 8509384 Storage Tube Plug Cap,Orange, for no. 850989168-528Log in pour la disponibilitéCase of 38,400 - $5,080.00
Informations sur le produit
- Catalog #AvailabilityPrice
- Empty racks with lids for 384 round or square storage tubes, Blue, for nos. 8509 and 8510Empty racks with lids for 384 round or square storage tubes, Blue, for nos. 8509 and 851089168-532Log in pour la disponibilitéCase of 20 - $366.00