Corning® Bar Coded Storage Tubes, Corning
96× 1D/2D barcoded storage tubes, with screw caps, Orange, 1.3 ml, bulk packed
: Corning
: 8502
PP. The 1D/2D storage line is an innovative product with both a linear 1D and 2D barcodes etched on each tube.
- Maximum information, 14×14 dot 2D barcode
- Barcode stability, laser-etched, not an attached label
- Compound compatibility, polypropylene construction throughout, making it inert
- Volume:1.3 ml
- Désignation:96× 1D/2D barcoded storage tubes, with screw caps, Orange
- Packaging:bulk packed
Accessories
- Description
- 89168-51496 screw caps for storage tubes 1,3 ml, with O-Ring, Orange, for no. 8500Case of 4,800 - $5,060.00
- Description
- 89168-51696 storage tube screw cap, with O-Ring, on mat, clear with cover, Orange, for no. 8500Case of 4,800 - $5,880.00
- Description
- 89168-518Thermoplastic elastomer (TPE) 96 storage tube septum cap, White, for no. 8500Case of 9,600 - $901.00
- Description
- 89168-520Thermoplastic elastomer (TPE) 96 storage tube septum cap on mat, White, for no. 8500Case of 4,800 - $1,480.00
- Description
- 89168-522Empty racks with lids for 96 screw capped storage tubes no. 8502, NaturalCase of 10 - $145.00
- Description
- 89168-530384 storage tube plug cap on mats for round tube no. 8509, 384 caps per mat, OrangeCase of 38,400 - $7,100.00
- Description
- Description
- 89168-532Empty racks with lids for 384 round or square storage tubes, Blue, for nos. 8509 and 8510Case of 20 - $366.00