Insulating Jackets for Jacketed Cylindrical Reactors, Chemglass | Avantor
Insulating Jackets for Jacketed Cylindrical Reactors, Chemglass
50 L, Insulating Jacket (Short)
Catalog # 89166-550
Supplier:   Chemglass
Fournisseur #:  CG-10011-120
Insulating Jackets for Jacketed Cylindrical Reactors, Chemglass
Catalog # 89166-550
Supplier:  Chemglass
Insulating Jackets for Jacketed Cylindrical Reactors, Chemglass
Insulating Jackets for Jacketed Cylindrical Reactors, Chemglass
50 L, Insulating Jacket (Short)
Price
$7,859.69
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
Insulating jacket is made from a heavy fiberglass fabric on the interior with a silicone-coated fiberglass fabric on the exterior.
  • Jacket is slit lengthwise to a hole in the bottom center
  • With lacing closure to accommodate a bottom drain valve
For use as a heat or cold shield on chemglass jacketed reactors.
Spécifications
  • Capacité:
    50 L
  • Désignation:
    Insulating Jacket (Short)