AUDA ≥98% (par CCM)
50 mg
Catalog # 89158-260
Supplier: Enzo Life Sciences
Fournisseur #: BML-EI392-0050
Jump to:
Product Details & Documents
Novel inhibitor of soluble epoxide hydrolase (IC50=18nM and 69nM for the mouse and human enzymes respectively). Produces a hypotensive effect in angiotensin-infused rats and prevents and reverses cardiac hypertrophy in mouse models of cardiac hypertrophy.
- Epoxide hydrolase inhibitor
White solid.
Soluble in DMSO (10mg/ml) or 100% ethanol (4mg/ml).
Synonymes: 12-(3-Adamantan-1-yl-ureido)-dodecanoic acid, 12-(3-(Adamantan-1-yl)ureido)dodecanoic acid
- CAS Number:479413-70-2
- Formule:C₂₁H₃₆N₂O₃
- Numéro MDL:MFCD12912267
- Molecular Weight:392.575 g/mol
- Température de conservation:Congélateur
Spécifications
Spécifications
- Taille:50 mg
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



