AUDA ≥98% (par CCM)
10 mg
Catalog # 89158-258
Jump to:
Discontinued
Product Details & Documents
Novel inhibitor of soluble epoxide hydrolase (IC50=18nM and 69nM for the mouse and human enzymes respectively). Produces a hypotensive effect in angiotensin-infused rats and prevents and reverses cardiac hypertrophy in mouse models of cardiac hypertrophy.
- Epoxide hydrolase inhibitor
White solid.
Soluble in DMSO (10mg/ml) or 100% ethanol (4mg/ml).
Synonymes: 12-(3-Adamantan-1-yl-ureido)-dodecanoic acid, 12-(3-(Adamantan-1-yl)ureido)dodecanoic acid
- CAS Number:479413-70-2
- Formule:C₂₁H₃₆N₂O₃
- Numéro MDL:MFCD12912267
- Molecular Weight:392.575 g/mol
- Température de conservation:Congélateur
Spécifications
Spécifications
- Taille:10 mg



