Back-Loading Printed Press-on Envelope, Associated Bag
Back-Loading Printed Press-on Envelope, Associated Bag
Back-loading printed press-on envelope, 6", Orange tint
Catalog # 89140-080
Supplier:   Associated Bag
Supplier #:  45-2-14
Back-Loading Printed Press-on Envelope, Associated Bag
Catalog # 89140-080
Supplier:  Associated Bag
Back-Loading Printed Press-on Envelope, Associated Bag
Back-Loading Printed Press-on Envelope, Associated Bag
Back-loading printed press-on envelope, 6", Orange tint
Price
$284.62
Log in for availability
Case of 1,000
Product Details & Documents
This envelope sticks to packages to protect shipping documents from dirt and moisture.
Back-loading style forms tamper-evident pocket for papers
Pressure-Sensitive Adhesive
Dimension-Listed Open End
"Packing List Enclosed" Printed on Envelope
Tamper-Evident Pocket
The pocket must be torn or cut open to retrieve papers.
Pressure-sensitive adhesive sticks aggressively to paper, metal, wood, corrugated cardboard.
Specifications
  • Color:
    Orange tint
  • Désignation:
    Back-loading printed press-on envelope
  • Dimensions:
    6"