Colorimetric and Fluorogenic Enzyme Substrates, Biotium
Beta-Glucuronidase Substrate Sampler Kit, 1 kit
Catalog # 89138-244
Supplier: Biotium
Fournisseur #: 10025
Colorimetric and Fluorogenic Enzyme Substrates, Biotium
Catalog # 89138-244
Supplier: Biotium
CAS Number:
Jump to:
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
Popular chromogenic and fluorogenic substrates for phosphatases, peroxidases, and other enzymes.
- Wide selection of popular substrates for alkaline phosphatase, horseradish peroxidase, beta-galactosidase, and other common enzymes
- Chromogenic substrates for absorbance-based detection or light microscopy
- Fluorogenic substrates for highly sensitive fluorescence-based ELISA
- Substrate sampler kits available
Biotium offers a number of popular substrates for alkaline phosphatase, beta-galactosidase, and horseradish peroxidase which are widely used for chromogenic or fluorogenic ELISA, immunohistochemistry, or blotting applications.
Beta-glucuronidase substrates are widely used to detect E. coli bacterial contamination in food and water, and also in reporter gene assays in plants.
X-beta-D-cellobioside is a substrate for beta-cellobiosidase.
ABTS is a registered trademark of Roche. Amplex Red is a registered trademark of Thermo Fisher Scientific.
Spécifications
Spécifications
- Désignation:Beta-Glucuronidase Substrate Sampler Kit
- Taille:1 kit