Colorimetric and Fluorogenic Enzyme Substrates, Biotium
Colorimetric and Fluorogenic Enzyme Substrates, Biotium
Purple-Beta-D-Gal, 25 mg
  89138-226
 :   Biotium
Supplier #:  10014
Colorimetric and Fluorogenic Enzyme Substrates, Biotium
  89138-226
 :  Biotium
 :  
Colorimetric and Fluorogenic Enzyme Substrates, Biotium
Purple-Beta-D-Gal, 25 mg
 :  
$271.47
Each (25mg)
Popular chromogenic and fluorogenic substrates for phosphatases, peroxidases, and other enzymes.
  • Wide selection of popular substrates for alkaline phosphatase, horseradish peroxidase, beta-galactosidase, and other common enzymes
  • Chromogenic substrates for absorbance-based detection or light microscopy
  • Fluorogenic substrates for highly sensitive fluorescence-based ELISA
  • Substrate sampler kits available
Biotium offers a number of popular substrates for alkaline phosphatase, beta-galactosidase, and horseradish peroxidase which are widely used for chromogenic or fluorogenic ELISA, immunohistochemistry, or blotting applications.
Beta-glucuronidase substrates are widely used to detect E. coli bacterial contamination in food and water, and also in reporter gene assays in plants.
X-beta-D-cellobioside is a substrate for beta-cellobiosidase.
ABTS is a registered trademark of Roche. Amplex Red is a registered trademark of Thermo Fisher Scientific.
  • Désignation:
    Purple-Beta-D-Gal
  • Taille:
    25 mg